MSA™ Ultra-Twin™ Respirators

Turned-in edge for a tight seal against the face and large chin cup for easy positioning

Manufacturer: MSA™ 480267

 View more versions of this product

Catalog No. 18-917

  Check Availability
Add to Cart

Required Accessories : Comfo Series cartridges and filters (see separate listings).

    Specifications

    Certifications NIOSH
    Color Black
    For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
    Includes Nosecup and speaking diaphragm
    Material (Lens) Polycarbonate
    Material Silicone
    Size Large
    Product Type Ultra-Twin™ Full Facepiece Respirator

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit