Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Inhalation Valve Seat

Catalog No. 189994099 Shop All MSA Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-999-4099 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-4099 Supplier MSA™ Supplier No. 459711
Add to Cart
Add to Cart

For use with Ultra-Twin™ respirators.

  • Allows air to enter the facepiece through the filtering media but prevents exhaled air from leaving the facepiece through the intake openings
  • For use with Ultra-Twin™ full-facepiece respirators
  • Sold in single package
  • MSA Model #459711

Specifications

Color Black
Product Type Inhalation Valve Seat
For Use With (Equipment) Ultra-Twin™ Respirators
Model 459711
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.