Learn More
MSA™ Ultralight™ Cap Lamp System: Chargers
Catalog No. 19047205 Shop All MSA Products
Change view
Click to view available options
Product Type:
5-unit AC Battery Charger
Single-unit AC Battery Charger
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-047-205 5-unit AC Battery Charger
19-047-203 Single-unit AC Battery Charger
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-047-205 Supplier MSA™ Supplier No. 10002460
Battery chargers for Ultralight Cap Lamp System
Single- or five-unit AC chargers. Five-unit charger can be internally set for 110V or 220V, mounted to a table or wall, or stacked to convert it to a multi-station charger for up to 60 stations. All charger stations are self-contained and microprocessor-controlled, requiring no calibration, adjustments, or rectifiers. LED lights indicate condition of battery at all times.
Order Info
Replacement batteries and clips for connecting Luminator battery to lamp top (two required) sold separately. Additional replacement parts are also available; contact you Fisher Scientific™ Customer Service Representative.
Specifications
Product Type 5-unit AC Battery Charger
Certifications/Compliance MSHA
For Use With (Equipment) Ultralight cap lamp system
Voltage 110/220 V