Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultravue™ Series Full-Facepiece Assembly

Catalog No. 19003375 Shop All MSA Products
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-003-375 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-003-375 Supplier MSA™ Supplier No. 801173
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pressure-demand NFPA facepiece made of hycar rubber provides comfort and easy fitting adjustment.

  • NFPA facepiece constructed of hycar rubber
  • Easy fit while providing rapid donning as well as simplified method of adjustment
  • Lens is scratch and chemical resistant
  • Five suspension head straps and roller buckles to secure in place

Specifications

Certifications/Compliance NIOSH
Color Black
Product Type Facepiece Assembly
For Use With (Equipment) Belt mounted SCBA
Material Hycar
Size Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.