Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultravue™ Silicone Full-Facepiece Respirator

Catalog No. 19076969 Shop All MSA Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-076-969 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-076-969 Supplier MSA™ Supplier No. 480255
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accessories for full-facepiece respirators

MSA Ultravue Full-Facepiece Respirators

  • Model 19-076-969 (Large)
  • Specifications

    Color Black
    Material Silicone
    Product Type Facepiece Assembly
    Format Large
    For Use With (Application) Protection against particulate matter, vapors and gas
    For Use With (Equipment) OptiMair™ 6A PAPR
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.