Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-Gard™ Chin Protectors

Catalog No. 17200042 Shop All MSA Products
Change view
Click to view available options
Product Type:
Retractable Chin Protector
Standard Chin Protector
Height (English):
2.86 in.
3.82 in.
Height (Metric):
7.2 cm
9.7 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Height (English) Height (Metric)
17-200-042 Retractable Chin Protector 3.82 in. 9.7 cm
17-200-041 Standard Chin Protector 2.86 in. 7.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-200-042 Supplier MSA™ Supplier No. 10115828
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provide extra protection from impact and splash hazards to face and neck

  • For use with select V-Gard Nitrometers
  • Opaque material ensures safety and standards compliance with clear, tinted, shaded and coated V-Gard visors
  • Compact, lightweight standard protector provides additional protection between and neck area
  • Retractable protector allows greater flexibility of head movement

Specifications

Certifications/Compliance ANSI Z87.1 2010-IAC-03-005-CAL
Color Black
For Use With (Equipment) V-Gard Nitrometer Visor
Height (English) 3.82 in.
Height (Metric) 9.7 cm
Material Polycarbonate
Product Type Retractable Chin Protector
Style Opaque compact
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.