Learn More
MSA™ V-Gard™ Replacement Polycarbonate Visors
Catalog No. 17200037 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
V-Gard Frame and Geadgear
V-Gard Retractable or Standard Chin Protector
Product Type:
V-Gard Visor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
17-200-037 V-Gard Retractable or Standard Chin Protector -
17-200-033 V-Gard Frame and Geadgear V-Gard Visor
17-200-038 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-200-037 Supplier MSA™ Supplier No. 10117783
Add to Cart
Add to Cart
Help protect wearers from flying debris, splash or spray
For use with V-Gard frames only; fit on all MSA Helmets.
  • Clear polycarbonate offers maximum light transmission and can be used indoors, outdoors and in low-light conditions
  • Maximum UV protection
  • Nitrometer style can be used only with V-Gard retractable or standard chin protector
Compliance
All visors meet EN 166, ANSI/ISEA Z87.1-2010 and AS/NZS 1337. 17-200-033 also meets CSA Z94.3.
Specifications
For Use With (Equipment) V-Gard Retractable or Standard Chin Protector