Learn More
MSA™ V-Gard™ Replacement Steel Mesh Visors
Catalog No. 17200039 Shop All MSA Products
Do not use where splash, elevated temperatures, UV or impact hazards exist
Catalog No. 17-200-039 Supplier MSA™ Supplier No. 10116557
Add to Cart
Add to Cart
Do not use where splash, elevated temperatures, UV or impact hazards exist
Provide maximum ventilation in humid working conditions
  • Lightweight and durable — great for outdoor applications
  • Epoxy-coated 20 x 20 mesh bound with heavy-duty plastic
  • Nitrometer style can be used only with V-Gard retractable of standard chin protector
Compliance
Meet EN 1731-F, ANSI/ISEA Z87.1-2010 (general protector) and AS/NZS 1337
Specifications
For Use With (Equipment) V-Gard Frame and Geadgear