Learn More
MSA™ V-Gard™ Slotted Protective Cap with 1-Touch Suspension
Catalog No. 191303445 Shop All MSA Products
Change view
Click to view available options
Color:
White
Yellow, Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-130-3445 White
19-130-3437 Yellow, Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Not specifically designed to protect against lateral blows from the front, side or rear.
Catalog No. 19-130-3445 Supplier MSA™ Supplier No. 10057441
Add to Cart
Add to Cart
Not specifically designed to protect against lateral blows from the front, side or rear.
Lightweight for all-day comfort
  • Polyethylene shell and suspension system work together as a protection system
  • Available in three sizes: small (6 to 71/8), standard (61/2 to 8), and large (71/2 to 81/2)
  • 1-Touch suspension allows one-handed slide adjustment and easy squeeze opening
Order Info
Contact your Fisher Scientific™ Customer Safety Representative for ordering information for sizes not shown.
Certifications
SEI certified.
Compliance
Meets or exceeds all applicable requirements of ANSI Z89.1-2003 for Type I helmets, Class E and CSA Z94.1 2005,
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2003; Type I; Class E and CSA Z94.1 2005; SEI certified
Color White
Material Polyethylene
Size Standard
Suspension Type 1-Touch
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.