Learn More
MSA™ V-Gard™ Visor for Chemical and Splash Applications
Catalog No. 17200199 Shop All MSA Products
Change view
Click to view available options
V-Gard Visor
Catalog No.
17-200-199 V-Gard Visor
1 options
Catalog No. 17-200-199 Supplier MSA™ Supplier No. 10115855
Add to Cart
Add to Cart
Propionate visor molded for superior optical quality
  • Manufactured with hand-polished molds for unsurpassed, distortion-free optical clarity
  • Clear visor with impact-rated protection
  • Extra thick to help prevent warping, cracking or crazing under tough work conditions
  • For use in manufacturing plants, laboratories and other industrial applications that demand face protection against impact hazards, abrasive particles, splash and spray
  • Meets requirements for EN 166 (MSA 1 B 3), ANSI/ISEA Z87.1-2010, CSA Z94.3 and AS/NZS 1337
  • Dimensions: 20.3 x 43.2 x 0.25cm (8 x 17 x 0.098 in.)
Specifications
Dimensions (L x W x H) 20.3 x 43.2 x 0.25 cm (8 x 17 x 0.098 in.)
For Use With (Application) Chemical and splash applications
Product Type V-Gard Visor