Learn More
MSA™ V-Gard™ Visor PC for General Purpose
Catalog No. 17200165 Shop All MSA Products
Change view
Click to view available options
Visor PC for General Purpose
Catalog No.
17-200-165 Visor PC for General Purpose
1 options
Catalog No. 17-200-165 Supplier MSA™ Supplier No. 10115836
Designed for use with V-Gard Frames and V-Gard Headgear
  • Polycarbonate visor protect against impact, chemical splash and UltraViolet radiation
  • Sides contoured towards face for closer fit and improved resistance to impact and splash hazards
  • Can be worn with helmet-mounted ear muffs
  • High UV protection
  • Available with Antifog/Antiscratch coatings
  • L x W x H: 20.3 x 43.1 x 0.10cm (8 x 17 x 0.04 in.)
Applications:
Sanding and Grinding, Food Processes, Painting, Asbestos Abatement
Specifications
Dimensions (L x W x H) 20.3 x 43.1 x 0.10 cm (8 x 17 x 0.04 in.)
For Use With (Equipment) V-Gard Frames and V-Gard Headgear
Product Type Visor PC