Learn More
MSA™ Workman™ Self Retracting Lanyards
Catalog No. 17200101 Shop All MSA Products
Catalog No. 17-200-101 Supplier MSA™ Supplier No. 10119507
Add to Cart
Add to Cart
  • RFID-enabled product for simplified product tracking and inspection
  • Integral, ergonomic carrying handle for easier transport and installation
  • 400 lb. working capacity for increased versatility
  • Swivel 36CS snaphook with load indicator at lifeline end greatly limits twisting of cable; also alerts users that unit has been involved in a fall
  • 30 ft. length for extra mobility
Certifications
CSA Z259.2.2 certified
Compliance
Meets OSHA 1926.502, 1910.66 and ANSI Z359.1-2007
Specifications
Length (English) 30 ft.
Certifications/Compliance Meets ANSI Z359.1-2007 and OSHA 1926.502, 1910.66. CSA Z259.2.2 certified.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.