Learn More
MSA™ Workman™ Tripod Confined Space Entry Kit
Catalog No. 17200100 Shop All MSA Products
Change view
Click to view available options
Confined space entry kit; With Lynx rescuer
Catalog No.
17-200-100 Confined space entry kit; With Lynx rescuer
1 options
Catalog No. 17-200-100 Supplier MSA™ Supplier No. 10117244
Ideal for multiple confined space
Workman Tripod Confined Space Entry Kit includes:
  • Lynx™ Rescuer allows for nonentry rescue of a confined space entrant
  • Lynx Hoist designed for lifting, lowering and positioning of personnel and materials
  • Carrying bags allow for greater portability
Order Info
Maximum Personnel Capacity: 400 lb. (182kg). Maximum Material Capacity: 620 lb. (280kg). Minimum Vertical Static Strength: 5000 lb. (22kN).
Compliance
Meets ANSI™ Z359.1, EN 795, and OSHA regulations.
Specifications
Includes Lynx rescuer
Strength 5000 lbf.
Load Bearing Capacity (Metric) 281 kg
For Use With (Application) Ideal for multiple confined space
Certifications/Compliance ANSI Z359.1, EN 795, OSHA
Load Bearing Capacity (English) 620 lb.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.