Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MSC 1186
GREENER_CHOICE

Catalog No. 792710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-271-0 10 mg
79-275-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 79-271-0 Supplier Tocris Bioscience™ Supplier No. 7927/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective SRPK family inhibitor

MSC1186 is a potent and selective inhibitor of serine arginine protein kinases (SRPK) 3, 1 and 2 (IC50 values are 0.6 nM, 3 nM and 81 nM respectively in biochemical assay; EC50 values in NanoBRETassay are 40 nM, 98 nM and 149 nM, respectively). Shows no activity at CLK and DYRK families (IC50 >10 μM).

Specifications

Chemical Name or Material N-[3-[[[2-(6-Chloro-7-fluoro-1H-benzimidazol-2-yl)-4-pyrimidinyl]amino]methyl]-2-pyridinyl]-N-methylmethanesulfonamide
CAS 2871698-23-4
Quantity 10 mg
Target SRPK3/STK23
Molecular Formula C19H17ClFN7O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.