Learn More
Selleck Chemical LLC MSC-4381-E1751-1G
SDP
Supplier:  Selleck Chemical LLC E17511G
Encompass_Preferred
MSC-4381 (MCT4-IN-1) is an orally active selective inhibitor of monocarboxylate transporter 4 (MCT4) with an IC50 of 77 nM and a Ki of 11 nM It has the potential to inhibit lactate efflux and promote intracellular lactate accumulation in MCT4-expressing cell lines
Catalog No. 50-313-7185
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.