Learn More
STEMCELL Technologies MSC Characterization Antibody Panel, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 1000354
Encompass
The MSC Characterization Antibody Panel is designed for the phenotypic analysis of human mesenchymal stromal cells (MSCs). This kit contains three fluorochrome-conjugated antibodies for the identification of human MSCs and a fluorochrome-conjugated antibody for the identification of non-MSCs. When measured by flow cytometry ≥ 95% of the MSC population must express CD73 CD90 and CD105 and these cells must lack expression ( 2% positive) of CD45. These reagents provide single-step labeling for the verification of human MSCs.The MSC Characterization Antibody Panel can be used to identify human MSCs derived from various tissues such as bone marrow MSCs pluripotent stem cell-derived mesenchymal progenitor cells adipose-derived MSCs dental pulp-derived MSCs and umbilical cord-derived MSCs.
Catalog No. 50-239-0350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.