Learn More
Promocell GmbH PromoCell Mesenchymal Stem Cell Chondrogenic Diff. Medium w/o Inducers 100 ml, ready-to-use, w/o Inducers with C-39814 attached to flask
Supplier: Promocell GmbH C28014
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).PromoCell offers five MSC Differentiation Media to efficiently induce differentiation of MSC into adipogenic, chondrogenic (w/ and w/o inducers), osteogenic or neurogenic lineages, respectively.PromoCell Mesenchymal Stem Cell Chondrogenic Differentiation Media is a serum-free medium developed for the directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into chondrogenic lineages.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.