Learn More
TARGETMOL CHEMICALS INC MSDC-0602K 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T38715
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MSDC-0602K (Azemiglitazone potassium) (Azemiglitazone potassium) is a PPAR(gamma)-sparing thiazolidinedione (Ps-TZD) compound that binds to PPAR(gamma) with an IC50 of 18.25 uM. It also modulates the mitochondrial pyruvate carrier (MPC). This compound MSDC-0602K has potential applications in researching fatty liver conditions including dysfunctional lipid metabolism inflammation and insulin resistance. MSDC-0602K acts as an insulin sensitizer improving insulinemia and fatty liver disease in mice both individually and in combination with Liraglutide. purity: 99%
Catalog No. 50-258-0091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.