Learn More
United States Biological Corporation Mouse Anti-Phosphatase, Alkaline, Liver, Bone, Kidney (Akp2, ALPI, ALPL) 100ug
Supplier:  United States Biological Corporation P407118100UG
Encompass_Preferred
Undifferentiated human Embryonal Carcinoma (EC) and Embryonic Stem (ES) cells have been shown to express very high levels of Alkaline Phosphatase isozyme that is indistinguishable from the isozyme found in liver, bone and kidney. Expression levels of AP decrease following stem cell differentiation. This antibody can be used to monitor the expression of the human Liver/Bone/Kidney isozyme of Alkaline Phosphatase (AP), and hence the differentiation status of human EC and ES cells by Flow Cytometry.
Catalog No. NC3083946
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.