Learn More
Genscript Corporation [Nle4, D-Phe7]-α-Melanocyte Stimulating Hormone (MSH), amide
Supplier:  Genscript Corporation RP106581
Encompass
[Nle4, DPhe7]-alpha-MSH (NDP-MSH), a highly potent analogue of alpha-melanocyte-stimulating hormone (alpha-MSH), possesses nanomolar efficacies at all the melanocortin receptor subtypes except the MC2R. Evaluation of the melanocortin 'message' sequence of [Nle4, DPhe7]-alpha-MSH was performed on the human melanocortin receptor subtypes designated hMC1, hMC3R, hMC4R, and hMC5R.
Catalog No. 50-274-922