Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501557650
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7650 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7650 Supplier Bethyl Laboratories, Inc Supplier No. A300451AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Germline mutations in human mismatch repair genes (hMSH2, hMSH6, hMLH1, hPMS2) account for majority of the hereditary non-polyposis colorectal carcinoma (HPNCC). CpG dinucleotides in the hMSH2 and hMLH1 genes are hotspots for HNPCC mutations. These mutations cause a mismatch repair deficiency resulting in a mutator phenotype where the replication errors are not repaired. Microsatellites/simple repeatative sequences are prone to this type of replication errors and instability of these microsatellites correlates with the occurance of HPNCC. hMSH2 binds to another MutS homolog protein GTBP to form a heterodimeric complex called hMutSbeta, which binds to insertion/deletion loops in DNA.

Specifications

Antigen MSH2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MSH2
Gene Accession No. P43246, P43247
Gene Alias COCA1, FCC1, HNPCC, HNPCC1
Gene Symbols MSH2
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17685, 4436
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.