Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551928 Supplier Invitrogen™ Supplier No. PA551928
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPEHSNTIDV CNTVDIKTED LSDSLPPVCD TVATDLCSTG IDICSFSEDI KPGDSLLLSV EEVLRSLETV SNTEVCCPNL QPNLEATVSN GPFLQLSSQS LSHNVFMSTS PALHGLSCTA ATPKIAKLNR KRSRSESDSE KVQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at lysine 16 which is implicated in the formation of higher-order chromatin structure. Acts as an E3 ubiquitin ligase that promotes monoubiquitination of histone H2B at 'Lys-35' (H2BK34Ub), but not that of H2A. This activity is greatly enhanced by heterodimerization with MSL1. H2B ubiquitination in turn stimulates histone H3 methylation at 'Lys-4' (H3K4me) and 'Lys-79' (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSL2
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MSL2
Gene Accession No. Q9HCI7
Gene Alias E130103E02Rik; E3 ubiquitin-protein ligase MSL2; E3 ubiquitin-protein transferase MSL2; Kiaa1585; male-specific lethal 2 homolog; male-specific lethal 2 homolog (Drosophila); male-specific lethal 2-like 1; Male-specific lethal-2 homolog; Male-specific lethal-2 homolog 1; MSL2; MSL-2; Msl2l1; MSL2-like 1; RGD1310355; ring finger protein 184; Rnf184
Gene Symbols MSL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MSL2. Recombinant protein control fragment (Product #RP-102003).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55167
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.