Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ msrB (Escherichia coli) Recombinant Protein

Catalog No. 89953072 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-072 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-072 Supplier Abnova™ Supplier No. P4999
Add to Cart
Add to Cart

Used for SDS-PAGE

Sequence: MGSSHHHHHHSSGLVPRGSHMANKPSAEELKKNLSEMQFYVTQNHGTEPPFTGRLLHNKRDGVYHCLICDAPLFHSQTKYDSGCGWPSFYEPVSEESIRYIKDLSHGMQRIEIRCGNCDAHLGHVFPDGPQPTGERYCVNSASLRFTDGENGEEING

Specifications

Accession Number NP_416292
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 947188
Molecular Weight (g/mol) 17.6kDa
Name msrB (Escherichia coli) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMANKPSAEELKKNLSEMQFYVTQNHGTEPPFTGRLLHNKRDGVYHCLICDAPLFHSQTKYDSGCGWPSFYEPVSEESIRYIKDLSHGMQRIEIRCGNCDAHLGHVFPDGPQPTGERYCVNSASLRFTDGENGEEING
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ECK1776/JW1767/yeaA
Common Name msrB
Gene Symbol msrB
Species E. coli
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.