Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent

Catalog No. PI48913 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Standard Components:
MSTFA
MSTFA + 1% TMCS
Quantity:
10 g
10 x 1 mL ampules
25 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Components Quantity
PI48913 MSTFA 25 mL
PI48911 MSTFA 10 g
PI48910 MSTFA 10 x 1 mL ampules
PI48915 MSTFA + 1% TMCS 10 x 1 mL ampules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI48913 Supplier Thermo Scientific™ Supplier No. TS48913
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.

Use Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent for GC analyses of early eluting compounds that would otherwise be obscured in the chromatogram. MSTFA, the most volatile TMS-amide available, and its byproduct N-methyltrifluoroacetamide typically elute with the solvent front, allowing TMS derivatives of small molecules to be analyzed. Addition of Thermo Scientific™ TMCS aids derivatization of amides, secondary amines, and hindered hydroxyls not derivatized by MSTFA alone.

  • Trimethylsilyl donor strength comparable to BSA and BSTFA
  • Reacts to replace labile hydrogens on a wide range of polar compounds with a Si(CH3)3 group
  • Used to prepare volatile and thermally stable derivatives for GC and MS
  • Volatile byproduct, N-methyltrifluoroacetamide, has an even lower retention time than MSTFA
  • Often TMS derivatives of small molecules can be analyzed when derivatized with MSTFA because the byproducts and the reagent itself usually elute with the solvent front
  • Addition of Thermo Scientfic TMCS aids derivatization of amides, secondary amines and hindered hydroxyls not derivatized by MSTFA alone
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC/MS
Hazards Flammable, Irritant
Storage 2°C to 8°C
Quantity 25 mL
Standard Components MSTFA
Standard Concentration 98 to 100%

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.