Learn More
Invitrogen™ MSX1/MSX2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MSX1/2.
Specifications
Specifications
| Antigen | MSX1/MSX2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MSX1 |
| Gene Accession No. | P13297, P28360, P35548, P52953, Q03358 |
| Gene Alias | AA675338; AI324650; BB122635; CRS2; ECTD3; FPP; homeo box, msh-like 1; homeo box, msh-like 2; homeobox 7; homeobox protein Hox-7; Homeobox protein Hox-8; homeobox protein Hox-8-1; Homeobox protein MSX-1; homeobox protein MSX-2; homeobox, msh-like 1; homeobox, msh-like 2; Hox7; Hox-7; Hox7.1; Hox-7.1; HOX8; Hox-8; Hox8.1; Hox-8.1; HYD1; msh; msh homeo box 1; msh homeo box 2; msh homeo box homolog 2; msh homeobox 1; msh homeobox 1-like protein; msh homeobox 2; msh homeobox homolog 1; msh homeobox homolog 2; Msx1; MSX2; Msx-2; muscle-segment homeobox; PFM; PFM1; STHAG1 |
| Gene Symbols | MSX1, MSX2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.