Learn More
IKA™ MT 40.10 Disposable Grinding Chamber
SDP
Catalog No. 01183083 Shop All IKA Works Products
Change view
Click to view available options
Blade Material:
Stainless Steel
Catalog No. Blade Material
01-183-083 Stainless Steel
1 options
This item is not returnable. View return policy
Catalog No. 01-183-083 Supplier IKA™ Supplier No. 4425000
Get a Magnetic Stirrer with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
IKA disposable grinding chamber with a stainless steel beater reduces soft, medium, hard and brittle materials with a Mohs hardness of up to 5 (manganese or apatite: 5 Mohs).
  • Useful volume: 40 mL
  • Quantity: 10 pieces/pack
  • Material grinding chamber: PP
  • Transparent plastic chamber so grinding tests can be observed at anytime
Specifications
Capacity (Metric) 40 mL
Blade Material Stainless Steel
Material Plastic
For Use With (Equipment) IKA Tube Mill Control, Tube Mill 100 Control
Model MT 40.10