Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT-ATP6, Mouse anti-Human, Clone: 1G7-1G2, Millipore Sigma™

Catalog No. MABS199525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS199525 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS199525 Supplier MilliporeSigma Supplier No. MABS199525UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MT-ATP6, clone 1G7-1G2, Cat. No. MABS1995, is a mouse monoclonal antibody that detects ATP synthase subunit a and has been tested for use in Western Blotting.

ATP synthase subunit a (UniProt: P00846; also known as F-ATPase protein 6) is encoded by the MT-ATP6 (also known as ATP6, ATPASE6, MTATP6) gene (Gene ID: 4508) in human. F-ATPase Protein 6 is an inner mitochondrial membrane protein and is a component of Complex V, which produces ATP from ADP in the presence of proton gradient across the membrane. F-type ATPases have 2 components, CF1 - the catalytic core - and CF0- the membrane proton channel. The CF1 catalytic core contains five subunits: alpha3, beta3, gamma1, delta1, epsilon1 and CF0 has three main subunits known as a, b, and c. Together they form a rotary motor. During catalysis, ATP synthesis in the catalytic domain of F1 is coupled via a rotary mechanism of the central stalk subunits to proton translocation. Defects in MT-ATP6 gene cause multiple mitochondrial diseases, such as Leber hereditary optic neuropathy (LHON), which is characterized by acute and subacute loss of central vision due to optic nerve dysfunction. Some defects also lead to mitochondrial complex V deficiency that leads to neuropathy, ataxia, and hypertrophic cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT-ATP6
Applications Western Blot
Classification Monoclonal
Clone 1G7-1G2
Conjugate Unconjugated
Dilution Western Blotting Analysis: 0.5μg/mL from a representative lot detected MT-ATP6 in mitochondria from human neonatal dermal fibroblasts and mitochondria from human neonatal dermal fibroblasts depleted of mtDNA (Courtesy of Michael F. Marusich, Ph.D., mAbDx, Inc., Eugene, OR USA).
Formulation Purified mouse monoclonal antibody IgG2b in HEPES-Buffered Saline (15 mM HEPES, 150 mM NaCl,  pH 7.2) with 0.02% sodium azide.
Gene Alias ATP synthase subunit a;F-ATPase protein 6
Gene Symbols MT-ATP6;ATP6;ATPASE6;MTATP6
Host Species Mouse
Immunogen A synthetic peptide corresponding to the N-terminus of human MT-ATP6.
Purification Method Protein L
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 1G7-1G2 detects ATP synthase subunit a (MT-ATP6) in mitochondria isolated from human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.