Learn More
Promega MTase-Glo Methyltransferase Assay, 400 assays
Catalog No. PRV7601 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
400 Assays
Catalog No. Quantity
PRV7601 400 Assays
1 options
Catalog No. PRV7601 Supplier Promega Supplier No. V7601
The MTase-Glo Assay is a bioluminescence-based assay that can be used to monitor the activities of methyltransferases (MTases) and their modulation by small molecules in a wide range of plate formats.
  • Measure Methyltransferase Activity Using a Universal Bioluminescent Assay
  • Simple add−and−read format
  • Use with any methyltransferase
  • No need to modify substrates
Specifications
Content And Storage <-65°C
Quantity 400 Assays