Learn More
Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent
Shop All Thermo Scientific Products
Description
Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent. MTBSTFA derivatizes hydroxyl, carboxyl, thiol and primary and secondary amines with typically >96% yields, and the reaction byproducts are volatile and neutral. TBDMS ethers are 104 times more stable to hydrolysis than the corresponding TMS ethers. Addition of Thermo Scientific™ TBDMCS increases silylation potential.
- Derivatives have a high molecular concentration at M-57.
Specifications
Specifications
| For Use With | GC |
| Hazards | Irritant |
| Storage | 2°C to 8°C |
| Quantity | 5 mL ampules |
| Standard Components | MTBSTFA |
| Standard Concentration | 98 to 100% |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.