Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent

Catalog No. PI48927 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Standard Components:
MTBSTFA
MTBSTFA + 1% TBMDS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Components
PI48927 MTBSTFA + 1% TBMDS
PI48920 MTBSTFA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI48927 Supplier Thermo Scientific™ Supplier No. TS48927
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent.

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent. MTBSTFA derivatizes hydroxyl, carboxyl, thiol and primary and secondary amines with typically >96% yields, and the reaction byproducts are volatile and neutral. TBDMS ethers are 104 times more stable to hydrolysis than the corresponding TMS ethers. Addition of Thermo Scientific™ TBDMCS increases silylation potential.

Additional Product Information:
  •  Derivatives have a high molecular concentration at M-57.
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Hazards Irritant
Storage 2°C to 8°C
Quantity 10 x 1 mL ampules
Standard Components MTBSTFA + 1% TBMDS
Standard Concentration 98 to 100, 0.1 to 1%

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.