Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MTEP hydrochloride
GREENER_CHOICE

Catalog No. p-200048367
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-211-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-211-0R Supplier Tocris Bioscience™ Supplier No. 2921/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective mGlu5 antagonist

MTEP hydrochloride is a potent, selective and non-competitive mGlu5 antagonist (IC50 = 5 nM in Ca2+-flux assay; Ki= 16 nM). Displays anxiolytic activity in vivo and is orally active.

Specifications

Chemical Name or Material 3-((2-Methyl-1,3-thiazol-4-yl)ethynyl)pyridine hydrochloride
CAS 1186195-60-7
Assay Percent Range >99%
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C11H8N2S.HCl
Formula Weight Observed MW: 236.72

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.