Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTERF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101935 Supplier Invitrogen™ Supplier No. PA5101935
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MTERF.

Transcription termination factor. Binds to a 28 bp region within the tRNA(Leu(uur)) gene at a position immediately adjacent to and downstream of the 16S rRNA gene; this region comprises a tridecamer sequence critical for directing accurate termination. Binds DNA along the major grove and promotes DNA bending and partial unwinding. Promotes base flipping. Transcription termination activity appears to be polarized with highest specificity for transcripts initiated on the light strand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTERF
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MTERF1
Gene Accession No. Q99551, Q9EPI8
Gene Alias 4931431L11Rik; 9230106K09Rik; mitochondrial transcription termination factor; mitochondrial transcription termination factor 1; Mitochondrial transcription termination factor 1a; mitochondrial transcription termination factor 1b; mTERF; Mterf1; mTERF1a; mTERF1b; transcription termination factor 1, mitochondrial; Transcription termination factor 1a, mitochondrial; transcription termination factor, mitochondrial
Gene Symbols MTERF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MTERF1(Accession Q99551), corresponding to amino acid residues N266-S316.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7978, 85261
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.