Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTF2 Polyclonal Antibody

Catalog No. PIPA535932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535932 Supplier Invitrogen™ Supplier No. PA535932
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (98%), Rat (96%), Chicken (90%).

Polycomb group (PcG) that specifically binds histone H3 trimethylated at 'Lys-36' (H3K36me3) and recruits the PRC2 complex. Acts by binding to H3K36me3, a mark for transcriptional activation, and recruiting the PRC2 complex, leading to enhance PRC2 H3K27me3 methylation activity. Regulates the transcriptional networks during embryonic stem cell self-renewal and differentiation. Promotes recruitment of the PRC2 complex to the inactive X chromosome in differentiating XX ES cells and PRC2 recruitment to target genes in undifferentiated ES cells. Required to repress Hox genes by enhancing H3K27me3 methylation of the PRC2 complex. In some conditions may act as an inhibitor of PRC2 activity: able to activate the CDKN2A gene and promote cellular senescence by suppressing the catalytic activity of the PRC2 complex locally. Binds to the metal-regulating-element (MRE) of MT1A gene promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTF2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Mtf2
Gene Accession No. Q9Y483
Gene Alias 9230112N11Rik; AA537621; C76717; dJ976O13.2; hPCl2; M96; Metal regulatory transcription factor 2; metal response element binding transcription factor 2; metal response element DNA-binding protein M96; metal response element-binding transcription factor 2; metal-response element DNA-binding protein M96; Metal-response element-binding transcription factor 2; mPCl2; MTF2; PCL2; polycomb-like 2; polycomb-like protein 2; putative DNA binding protein; RGD1304727; RP5-976O13.1; TDRD19A; tudor domain containing 19A; zinc-regulated factor 1; ZiRF1
Gene Symbols Mtf2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human MTF2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.