Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTFMT Monoclonal Antibody (OTI1C1)
GREENER_CHOICE

Catalog No. PIMA525682
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA525682 100 μL
1 options

Catalog No. PIMA525682

Supplier: Invitrogen™ MA525682

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MTFMT Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this nuclear gene localizes to the mitochondrion, where it catalyzes the formylation of methionyl-tRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTFMT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTFMT
Gene Accession No. Q96DP5
Gene Alias 2310020P08Rik; COXPD15; Fmt; FMT1; LOW QUALITY PROTEIN: methionyl-tRNA formyltransferase, mitochondrial; methionyl-tRNA formyltransferase, mitochondrial; mitochondrial methionyl-tRNA formyltransferase; mitochondrial methionyl-tRNA transformylase; MtFMT; RGD1309462
Gene Symbols MTFMT
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 175-389 of MTFMT produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123263
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.