Learn More
Invitrogen™ MTFR1L Polyclonal Antibody

Description
Immunogen sequence: SEERLLALKK PALPALSRTT ELQDELSHLR SQIAKIVAAD AASASLTPDF LSPGSSNVSS PLPCFGSSFH STTS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 99%.
Specifications
Specifications
| Antigen | MTFR1L |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MTFR1L |
| Gene Accession No. | Q9H019 |
| Gene Alias | FAM54B; family with sequence similarity 54 member B; family with sequence similarity 54, member B; HYST1888; mitochondrial fission regulator 1 like; mitochondrial fission regulator 1-like; MST116; MSTP116; MTFR1L; protein FAM54B |
| Gene Symbols | MTFR1L |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.