Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MTG16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597544
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597544 Supplier Invitrogen™ Supplier No. PA597544
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTG16 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the myeloid translocation gene family which interact with DNA-bound transcription factors and recruit a range of corepressors to facilitate transcriptional repression. The t(16;21)(q24;q22) translocation is one of the less common karyotypic abnormalities in acute myeloid leukemia. The translocation produces a chimeric gene made up of the 5'-region of the runt-related transcription factor 1 gene fused to the 3'-region of this gene. This gene is also a putative breast tumor suppressor. Alternative splicing results in transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTG16
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CBFA2T3
Gene Accession No. O75081
Gene Alias A630044F12Rik; AI465270; AW229127; CBFA2/RUNX1 partner transcriptional co-repressor 3; CBFA2/RUNX1 translocation partner 3; CBFA2T3; Cbfa2t3h; core-binding factor, runt domain, alpha subunit 2, translocated to, 3; core-binding factor, runt domain, alpha subunit 2, translocated to, 3 (human); core-binding factor, runt domain, alpha subunit 2; translocated to, 3; eight twenty one protein 2; ETO/MTG8-related protein ETO-2; Eto2; ETO-2; hMTG16; MTG16; MTG16HEL; MTG8-related gene 2; MTG8-related protein 2; MTGR2; myeloid translocation gene 8 and 16b; myeloid translocation gene on chromosome 16 protein; myeloid translocation gene-related protein 2; protein CBFA2T3; Protein ETO-2; RUNX1T3; translocated to, 3; zinc finger MYND domain-containing protein 4; ZMYND4
Gene Symbols CBFA2T3
Host Species Rabbit
Immunogen Recombinant Human Protein CBFA2T3 protein (39-159AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 863
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.