Learn More
Invitrogen™ MTHFD2L Polyclonal Antibody

Description
MTHFD2L Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | MTHFD2L |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | MTHFD2L |
| Gene Accession No. | D3YZG8, Q9H903 |
| Gene Alias | 1110019K23Rik; C630010D07Rik; FLJ13105; LOW QUALITY PROTEIN: probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; methylenetetrahydrofolate dehydrogenase 2-like; MGC45532; MGC72244; MTHFD2L; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like; putative bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like protein; RGD1310879; tetrahydrofolate dehydrogenase/cyclohydrolase; Unknown (protein for MGC:63479); wu:fi14c12; zgc:63479 |
| Gene Symbols | MTHFD2L |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.