Learn More
TARGETMOL CHEMICALS INC MTOR INHIBITOR 9D 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T67702
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. mTOR inhibitor 9d is a dual inhibitor of the protein kinases mTOR and PI3K with an mTOR IC50 value of 0.31 nm and can be used for the treatment of leukemia skin cancer breast cancer lung cancer and colon cancer. purity: 98%
Catalog No. 50-257-9283
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.