Learn More
Sigma Aldrich Fine Chemicals Biosciences mTRAQR Reagent Delta4 50 U
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 44276981KT
Encompass
Contains mTRAQ(R) Reagents Delta4 for 50 mTRAQ assays. One unit of reagent can label up to 100 mug of protein. Pack has 1 tube each of Delta4 containing 50 units of reagent. Optional Multiplex Buffer kit (Cat. No. 4381664) to be ordered separately.
Catalog No. 50-290-8046
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.