Learn More
Liofilchem™ MTS™ Cefoperazone*- Sulbactam (2/1) [CPS] 0.016-256* μg/mL

MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$51.25 - $418.00
Specifications
Antimicrobial Agents | Cefoperazone*-Sulbactam 2/1 |
---|---|
Concentration | 0.016 to 256* μg/mL |
Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
Description | Liofilchem™ MTS™ Cefoperazone*-Sulbactam 2/1 |
Antibiotic Code | CPS |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity & Availability | ||||
22-777-719
![]() |
Liofilchem™
920231 |
10/Pk. | Individually Packed |
Pack of 10 for $51.25
|
|
||||
22-777-717
![]() |
Liofilchem™
92023 |
30/Pk. | Individually Packed |
Pack of 30 for $133.00
|
|
||||
22-777-718
![]() |
Liofilchem™
920230 |
100/Pk. | Canister Pack |
Pack of 100 for $418.00
|
|
||||
Description
Specifications
Cefoperazone*-Sulbactam 2/1 | |
-20°C or 2°C to 8°C until the given expiry date | |
CPS | |
Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
0.016 to 256* μg/mL | |
Liofilchem™ MTS™ Cefoperazone*-Sulbactam 2/1 | |
MIC Test Strip |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.