Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liofilchem™ MTS™ Ceftazidime-Avibactam [CZA] 0.016/4-256/4 μg/mL
SDP

Catalog No. p-7146368 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
30/Pk.
Packaging Type:
Canister Pack
Individually Packed
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
22-777-941 10/Pk. Individually Packed
22-777-939 30/Pk. Individually Packed
22-777-940 100/Pk. Canister Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-777-941 Supplier Liofilchem™ Supplier No. 921391
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.

  • Ready to use. No preparation or dilution of antimicrobial agent is required. Simply place on an inoculated agar plate and incubate
  • MIC testing for a wide range of agents and against many organisms can be done on request and at short notice
  • Synergy Testing: can be crossed for evaluating a critical assay such as the in vitro combination of antibiotics
  • Rapid and Efficient: easy to set up. Full range of MIC testing of 6 agents on a large plate takes no more than 5 minutes
  • Solid Medium: Growth of organisms on agar better mimics in vitro conditions. It is easy to visualize hetero-resistant variants in a population and to detect mixed cultures and contamination
  • Inoculum Tolerant, Macro.Method: a stable gradient technique that is inoculum tolerant (changes in inoculum do not affect the MIC results for a susceptible strain), thus it can be used in a macro-format with heavier inoculum and a richer media to maximize the detection of low-level and hetero-resistance
  • Easy to Handle: The package is small and light weigh. Required storage space is minimal
  • Wide MIC Range: spans across 15 dilutions fulfilling pharmacokinetic needs, varying susceptibility of many organism groups, and covers different MIC breakpoints used in different countries as recommended by various reference or expert groups
  • Easy to Learn: Minimal training is required for clinical microbiology personnel
  • No equipment is required for use
  • Easy for New Agents
  • MIC Equivalent to Reference Values
  • Improved MIC Precision
  • Flexible Media Choice: can be used as a quality control tool for checking different AST media, brand to brand and batch to batch
  • Robust Technique
  • Generates quantitative MIC values that are needed to guide therapy selection for the individual patient - choice, dose and regimen
  • Can be used for Fastidious and Slow Growers
  • Can be used to directly test positive blood cultures, CSF and sputum from cystic fibrosis patients and bronchial aspirates from VAP patients. The extra day saved by testing the specimen directly can have significant impact in therapy management, patient care and hospital cost savings
  • Specifications

    Antimicrobial Agents Ceftazidime-Avibactam
    Certifications/Compliance FDA Cleared
    Concentration 0.016/4 to 256/4 μg/mL
    Content And Storage -20°C or 2°C to 8°C until the given expiry date
    Description Liofilchem™ MTS™ Ceftazidime-Avibactam
    Antibiotic Code CZA
    Product Type MIC Test Strip
    For Use With (Application) Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms
    Packaging Type Individually Packed
    Quantity 10/Pk.

    Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.