Learn More
Apexbio Technology LLC Colorimetric Nitric Oxide (NO) Assay Kit 2500T
SDP
Supplier:  Apexbio Technology LLC K21072500
Encompass_Preferred
The Colorimetric Nitric Oxide (NO) Assay Kit is designed for the quantitative detection of nitric oxide production in biological samples This assay operates by converting nitrate to nitrite via enzymatic reduction followed by the reaction of nitrite with Griess reagents to yield a chromophore measurable at 540 570 nm The method enables sensitive assessment of nitric oxide bioavailability facilitating studies on cellular signaling oxidative stress and inflammatory responses This kit is broadly applicable for research investigating nitric oxide-mediated physiological and pathological processes in various cell and tissue models
Catalog No. 50-000-97197