Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 2 Monoclonal Antibody (107-82)
GREENER_CHOICE

Catalog No. MA3942 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3942 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3942 Supplier Invitrogen™ Supplier No. MA3942
  • $600.50
    $449.65 / Each of 1

    Save $150.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-942 detects the 80 kDa subunit of m-calpain in human platelets and erythrocytes, bovine platelets, heart and skeletal muscle, rat myoblasts, kidney, liver and spleen, pig cultured cells, and rabbit samples. This antibody does not cross-react with mu-calpain, n-calpain, calmodulin or calpastatin. MA3-942 has been successfully used in Western blot, IF, immunohistochemistry, and ELISA procedures. By Western blot, this antibody detects an ~80 kDa protein representing m-calpain in bovine platelets. Immunocytochemical staining of m-calpain in LLC-PK1 cells with MA3-942 results in diffuse cytoplasmic staining. This product has not been shown to be effective in immunoprecipitation experiments. The MA3-942 antigen is purified bovine skeletal muscle 80 kDa subunit of m-calpain. This antibody recognizes an epitope between amino acids 502-699 (domain III/IV) of human calpain.

Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 107-82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene CAPN2
Gene Accession No. P06814, P17655, P43367, Q07009, Q27971
Gene Alias 80 kDa M-calpain subunit; AI326419; Calcium-activated neutral proteinase 2; calcium-dependent protease; CALP80; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; CANP2; CANPL2; CANPml; Capa2; Capa-2; CAPN2; cysteine proteinase; m-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; m-calpin; mCANP; millimolar-calpain
Gene Symbols CAPN2
Host Species Mouse
Immunogen Purified bovine skeletal muscle 80 kDa subunit of m-calpain.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009092, 281662, 29154, 397393, 824
Target Species Human, Rat, Rabbit, Bovine, Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.