Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MU-Gal, Beta-Galactosidase Substrate, >95% Purity, 1 g

Catalog No. 502704688
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4688 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4688 Supplier Abcam Supplier No. AB1463561G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent substrate for detecting β-galactosidase in cell extracts and purified enzyme preparations.

  • Molecular Weight: 338.31 g/mol
  • Purity: >95%
  • Application: Fluorescent substrate for detecting β-galactosidase in cell extracts and purified enzyme preparations
  • Detection Method: Fluorescence microplate reader or fluorometer
  • Product Notes: Yields a water-soluble blue fluorescent coumarin fluorophore upon cleavage
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Β-galactosidase
Substrate 4-Methylumbelliferyl-beta-D-galactopyranoside
Shipping Condition Ambient
For Use With (Equipment) Fluorescence Microplate Reader, Fluorometer
Buffer 100% 4-Methylumbelliferyl-beta-D-galactopyranoside
Detection Method Fluorescence
Content And Storage Store at -20°C.
Formulation 100% 4-Methylumbelliferyl-beta-D-galactopyranoside
Sample Type Cell extracts, Purified enzyme preparations
Quantity 1 g
For Use With (Application) Detection of β-galactosidase in cell extracts and purified enzyme preparations
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.