Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MU-GLU, Fluorogenic Beta-Glucosidase Substrate, 338.32 g/mol, 100 mg

Catalog No. 502704686
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4686 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4686 Supplier Abcam Supplier No. AB146358100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic substrate for characterizing β-glucosidases and evaluating β-glucocerebrosidase activity.

  • Molecular weight: 338.32 g/mol
  • Applications: Fluorogenic substrate for characterizing β-glucosidase and β-glucocerebrosidase; used in assays to evaluate β-glucocerebrosidase deficiency related to Gaucher disease
  • Fluorescence: λex 365 nm; λem 445 nm in 0.1 M sodium acetate pH 5.0 (after cleavage by β-glucosidase)
  • Hydrolysis product: 4-MU
  • Composition: 100% MU-GLU

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Beta-Glucosidase, Beta-Glucocerebrosidase
Substrate Properties Chemical Substrate
Substrate MU-GLU
Shipping Condition Blue Ice
Buffer 100% MU-GLU
Content And Storage Store at -20°C.
Detection Method Fluorescence
Formulation 100% MU-GLU
Quantity 100 mg
For Use With (Application) Characterizing novel β-glucosidases, Gaucher disease assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.