Learn More
Selleck Chemical LLC Mubritinib (TAK 165) 10mM/1mL 366017-09-6
SDP
Supplier:  Selleck Chemical LLC S221610MM/1ML
Encompass_Preferred
Mubritinib (TAK-165) is a potent inhibitor of HER2/ErbB2 with IC50 of 6 nM in BT-474 cell; no activity to EGFR, FGFR, PDGFR, JAK1, Src and Blk in BT-474 cell line. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.