Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci MUC-17 Recombinant Protein
SDP

Catalog No. 16833562
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-833-562 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-833-562 Supplier ProSci Supplier No. 111890100
Add to Cart
Add to Cart

Recombinant Protein

This protein carries a polyhistidine tag at the C-terminus. This protein was cleaved within the SEA domain between 4243 Gly and 4244 Ser, and was cleaved into N and C-terminal fragment with calculated MW of 12.6 kDa and 18.6 kDa respectively. The protein

Membrane mucins have several functions in epithelial cells including cytoprotection, extravasation during metastases, maintenance of luminal structure, and signal transduction. MUC17, contains an extended, repetitive extracellular glycosylation domain and a carboxyl terminus with two EGF-like domains, a SEA module domain, a transmembrane domain, and a cytoplasmic domain with potential serine and tyrosine phosphorylation sites. Interacts via its C-terminus with PDZK1 and this interaction appears important for proper localization. Probably plays a role in maintaining homeostasis on mucosal surfaces.

Specifications

For Use With (Application) Western Blot
Molecular Weight (g/mol) 31.2 kDa
Quantity 0.1 mg
Source Human Cells
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His Tag
Sequence Arg 4131 - Leu 4390
Content And Storage Lyophilized Protein should be stored at -20°C or lower for long term storage. Upon reconstitution, working aliquots should be stored at -20°C or-70°C. Avoid repeated freeze-thaw cycles.
Purity or Quality Grade >90% as determined by SDS-PAGE.
Synonym Mucin-17,MUC-17,MUC17,MUC-3,MUC3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.