Learn More
Description
C595 (NCRC48) recognizes CD227, also known as mucin 1 which is a breast cancer associated mucin encoded by the Muc-1 gene. Mucins are a family of high molecular weight, heavily glycosylated proteins (glycoconjugates) produced by many epithelial tissues in vertebrates. CD227 is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. This protein is overexpressed abundantly in >90% breast carcinomas and metastases.Mouse anti Human CD227 antibody, clone C595 recognizes the peptide epitope ARG-PRO-ALA-PRO within the protein core of the mucin.
Specifications
Specifications
| Antigen | MUC1 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | C595 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | MUC1 |
| Gene Accession No. | P15941 |
| Gene Alias | ADMCKD; ADMCKD1; Breast carcinoma-associated antigen DF3; CA 15-3; Cancer antigen 15-3; carcinoma-associated mucin; CD227; DF3 antigen; EMA; Episialin; H23 antigen; H23AG; J19; KL-6; Krebs von den Lungen-6; MAM6; MCD; MCKD; MCKD1; Medullary cystic kidney disease, autosomal dominant; Muc1; MUC-1; MUC-1/SEC; MUC-1/X; MUC1/ZD; MUC1-alpha; MUC1-beta; MUC1-CT; MUC1-NT; mucin 1, cell surface associated; mucin 1, transmembrane; Mucin1; mucin-1; Mucin-1 subunit alpha; Mucin-1 subunit beta; peanut-reactive urinary mucin; PEM; PEMT; Polymorphic epithelial mucin; PUM; tumor associated epithelial mucin; tumor-associated epithelial membrane antigen; Tumor-associated mucin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
