Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572931
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572931 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572931 Supplier Invitrogen™ Supplier No. PA572931
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MUC12 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), IP, ELISA

Epitheial mucins are secreted glycoproteins that play roles in cell protection, adhesion modulation and signaling. MUC12 is a novel mucin gene and may be involved in epithelial cell growth regulation. MUC12 is down-regulated in colorectal cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC12
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MUC12
Gene Accession No. Q9UKN1
Gene Alias MUC11; MUC-11; MUC12; MUC-12; mucin 11; mucin 12, cell surface associated; mucin-11; Mucin12; Mucin-12
Gene Symbols MUC12
Host Species Rabbit
Immunogen Residues 5434-5448 of the human MUC 12 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10071
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.