Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC2 Recombinant Rabbit Monoclonal Antibody (HL1724)
GREENER_CHOICE

Catalog No. PIMA547157 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547157 Supplier Invitrogen™ Supplier No. MA547157
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1724
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MUC2
Gene Accession No. Q02817, Q62635, Q80Z19
Gene Alias 2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn
Gene Symbols MUC2
Host Species Rabbit
Immunogen Synthtic peptide of human MUC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17831, 24572, 4583
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.